Assay Details
Gene Symbol: camsap2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044996.2 | NP_001038461.1 | 13 - 14 | 3596 | 85 | |
XM_009296260.3 | XP_009294535.1 | 14 - 15 | 4080 | 85 | ||
XM_009296261.3 | XP_009294536.1 | 13 - 14 | 4047 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
