Assay Details
Gene Symbol: camsap2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044996.2 | NP_001038461.1 | 16 - 17 | 3919 | 69 | |
XM_005170467.4 | XP_005170524.1 | 17 - 18 | 4379 | 69 | ||
XM_005170468.4 | XP_005170525.1 | 16 - 17 | 4346 | 69 | ||
XM_009296260.3 | XP_009294535.1 | 17 - 18 | 4403 | 69 | ||
XM_009296261.3 | XP_009294536.1 | 16 - 17 | 4370 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
