Assay Details
Gene Symbol: camsap2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044996.2 | NP_001038461.1 | 2 - 3 | 399 | 73 | |
XM_005170467.4 | XP_005170524.1 | 2 - 3 | 835 | 73 | ||
XM_005170468.4 | XP_005170525.1 | 2 - 3 | 835 | 73 | ||
XM_009296260.3 | XP_009294535.1 | 2 - 3 | 835 | 73 | ||
XM_009296261.3 | XP_009294536.1 | 2 - 3 | 835 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
