Assay Details
Gene Symbol: c1qtnf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080078.1 | NP_001073547.1 | 3 - 4 | 457 | 84 | |
XM_005162187.4 | XP_005162244.1 | 3 - 4 | 1485 | 84 | ||
GenBank mRNA | BC128849.1 | - | 3 - 4 | 457 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
