Assay Details
Gene Symbol: c1qtnf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080078.1 | NP_001073547.1 | 4 - 5 | 638 | 88 | |
XM_005162187.4 | XP_005162244.1 | 4 - 5 | 1666 | 88 | ||
GenBank mRNA | BC128849.1 | - | 4 - 5 | 638 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
