Assay Details
Gene Symbol: c1qtnf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080078.1 | NP_001073547.1 | 5 - 6 | 747 | 101 | |
XM_005162187.4 | XP_005162244.1 | 5 - 6 | 1775 | 101 | ||
GenBank mRNA | BC128849.1 | - | 5 - 6 | 747 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
