Assay Details
Gene Symbol: gc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109695.1 | NP_001103165.1 | 9 - 10 | 2131 | 99 | |
XM_017356806.2 | XP_017212295.1 | 9 - 10 | 2131 | 99 | ||
XM_021477384.1 | XP_021333059.1 | 10 - 11 | 2426 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
