Assay Details
Gene Symbol: pcbd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200814.1 | NP_957108.1 | 2 - 3 | 154 | 120 | |
GenBank mRNA | AF473825.1 | - | 1 - 2 | 135 | 120 | |
BC059694.1 | - | 2 - 3 | 154 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
