Assay Details
Gene Symbol: rnf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205553.1 | NP_991116.1 | 3 - 4 | 704 | 87 | |
NM_205766.1 | NP_991329.1 | 3 - 4 | 757 | 87 | ||
GenBank mRNA | BC044545.1 | - | 3 - 4 | 757 | 87 | |
BC065643.1 | - | 3 - 4 | 704 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
