Assay Details
Gene Symbol: gnmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212816.1 | NP_997981.1 | 4 - 5 | 670 | 75 | |
GenBank mRNA | BC062527.1 | - | 4 - 5 | 670 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
