Assay Details
Gene Symbol: naa10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213334.1 | NP_998499.1 | 5 - 6 | 460 | 122 | |
GenBank mRNA | BC053180.1 | - | 5 - 6 | 460 | 122 | |
BC165054.1 | - | 5 - 6 | 376 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
