Assay Details
Gene Symbol: rel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001841.3 | NP_001001841.2 | 7 - 8 | 747 | 73 | |
GenBank mRNA | BC076403.1 | - | 7 - 8 | 751 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
