Assay Details
Gene Symbol: rac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002061.1 | NP_001002061.1 | 3 - 4 | 438 | 84 | |
GenBank mRNA | AY865569.1 | - | 2 - 3 | 186 | 84 | |
BC071369.1 | - | 3 - 4 | 438 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
