Assay Details
Gene Symbol: rac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002061.1 | NP_001002061.1 | 4 - 5 | 499 | 74 | |
GenBank mRNA | AY865569.1 | - | 3 - 4 | 247 | 74 | |
BC071369.1 | - | 4 - 5 | 499 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
