Assay Details
Gene Symbol: rab3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002530.1 | NP_001002530.1 | 2 - 3 | 374 | 73 | |
XM_005166825.4 | XP_005166882.1 | 2 - 3 | 411 | 73 | ||
GenBank mRNA | BC076367.1 | - | 2 - 3 | 374 | 73 | |
BC165042.1 | - | 1 - 2 | 257 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
