Assay Details
Gene Symbol: uxt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004671.1 | NP_001004671.1 | 5 - 6 | 619 | 70 | |
GenBank mRNA | BC081425.1 | - | 5 - 6 | 619 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
