Assay Details
Gene Symbol: ifi30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006057.1 | NP_001006057.1 | 3 - 4 | 485 | 79 | |
GenBank mRNA | BC083267.1 | - | 3 - 4 | 485 | 79 | |
BC165122.1 | - | 3 - 4 | 422 | 79 | ||
JX018169.1 | - | 3 - 4 | 388 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
