Assay Details
Gene Symbol: rap2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007055.1 | NP_001007056.1 | 1 - 2 | 310 | 87 | |
GenBank mRNA | BC083208.1 | - | 1 - 2 | 310 | 87 | |
BC164968.1 | - | 1 - 2 | 308 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
