Assay Details
Gene Symbol: gpr137c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007433.1 | NP_001007434.1 | 1 - 2 | 685 | 93 | |
XM_005169819.4 | XP_005169876.1 | 1 - 2 | 724 | 93 | ||
GenBank mRNA | BC085410.1 | - | 1 - 2 | 685 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
