Assay Details
Gene Symbol: gpr137c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007433.1 | NP_001007434.1 | 6 - 7 | 1354 | 78 | |
GenBank mRNA | BC085410.1 | - | 6 - 7 | 1354 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
