Assay Details
Gene Symbol: pcyox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017626.1 | NP_001017626.1 | 4 - 5 | 648 | 66 | |
GenBank mRNA | BC092824.1 | - | 4 - 5 | 648 | 66 | |
BC164221.1 | - | 3 - 4 | 523 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
