Assay Details
Gene Symbol: acot9.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037399.1 | NP_001032476.1 | 13 - 14 | 1383 | 78 | |
GenBank mRNA | BC108006.1 | - | 13 - 14 | 1383 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
