Assay Details
Gene Symbol: tnfrsf21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042688.2 | NP_001036153.2 | 5 - 6 | 2005 | 80 | |
GenBank mRNA | BC141795.1 | - | 5 - 6 | 2005 | 80 | |
DQ812118.1 | - | 5 - 6 | 1676 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
