Assay Details
Gene Symbol: enah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013521.2 | NP_001013539.2 | 10 - 11 | 1724 | 71 | |
NM_001045028.3 | NP_001038493.1 | 11 - 12 | 2461 | 71 | ||
XM_009294716.3 | XP_009292991.1 | 10 - 11 | 1779 | 71 | ||
XM_021468415.1 | XP_021324090.1 | 11 - 12 | 1885 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
