Assay Details
Gene Symbol: enah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013521.2 | NP_001013539.2 | 11 - 12 | 1807 | 115 | |
NM_001045028.3 | NP_001038493.1 | 12 - 13 | 2544 | 115 | ||
XM_009294710.3 | XP_009292985.1 | 13 - 14 | 2659 | 115 | ||
XM_009294712.3 | XP_009292987.1 | 13 - 14 | 2428 | 115 | ||
XM_009294713.3 | XP_009292988.1 | 13 - 14 | 2611 | 115 | ||
XM_009294714.3 | XP_009292989.1 | 11 - 12 | 2242 | 115 | ||
XM_009294715.3 | XP_009292990.1 | 12 - 13 | 1923 | 115 | ||
XM_009294716.3 | XP_009292991.1 | 11 - 12 | 1862 | 115 | ||
XM_021468414.1 | XP_021324089.1 | 13 - 14 | 2029 | 115 | ||
XM_021468415.1 | XP_021324090.1 | 12 - 13 | 1968 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
