Assay Details
Gene Symbol: isl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130962.1 | NP_571037.1 | 5 - 6 | 1140 | 57 | |
XM_005165264.4 | XP_005165321.1 | 5 - 6 | 1175 | 57 | ||
XM_021475997.1 | XP_021331672.1 | 5 - 6 | 1181 | 57 | ||
GenBank mRNA | BC060892.1 | - | 5 - 6 | 1104 | 57 | |
D21135.1 | - | 5 - 6 | 1140 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
