Assay Details
Gene Symbol: glra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131402.1 | NP_571477.1 | 6 - 7 | 772 | 65 | |
XM_005173095.4 | XP_005173152.1 | 8 - 9 | 1584 | 65 | ||
XM_021480963.1 | XP_021336638.1 | 7 - 8 | 1578 | 65 | ||
GenBank mRNA | AF488379.1 | - | 8 - 9 | 797 | 65 | |
AJ005812.1 | - | 6 - 7 | 772 | 65 | ||
BC162943.1 | - | 6 - 7 | 740 | 65 | ||
BC162956.1 | - | 6 - 7 | 740 | 65 | ||
KM655639.1 | - | 6 - 7 | 680 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
