Assay Details
Gene Symbol: stat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131479.1 | NP_571554.1 | 16 - 17 | 1795 | 58 | |
XM_005163793.4 | XP_005163850.2 | 16 - 17 | 1689 | 58 | ||
XM_005163794.4 | XP_005163851.2 | 16 - 17 | 1689 | 58 | ||
XM_005163795.4 | XP_005163852.2 | 16 - 17 | 1689 | 58 | ||
XM_005163796.3 | XP_005163853.1 | 16 - 17 | 1795 | 58 | ||
XM_021469347.1 | XP_021325022.1 | 16 - 17 | 1795 | 58 | ||
GenBank mRNA | AB018219.1 | - | 16 - 17 | 1771 | 58 | |
AJ005693.1 | - | 16 - 17 | 1795 | 58 | ||
BC045276.1 | - | 15 - 16 | 1732 | 58 | ||
BC068320.1 | - | 16 - 17 | 1756 | 58 | ||
KU559611.1 | - | 16 - 17 | 1795 | 58 | ||
KU559612.1 | - | 16 - 17 | 1795 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
