Assay Details
Gene Symbol: nfat5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199895.1 | NP_956189.1 | 10 - 11 | 1859 | 117 | |
GenBank mRNA | BC058868.1 | - | 10 - 11 | 1859 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
