Assay Details
Gene Symbol: bcap31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200092.1 | NP_956386.1 | 4 - 5 | 450 | 87 | |
GenBank mRNA | BC049014.1 | - | 4 - 5 | 450 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
