Assay Details
Gene Symbol: sp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200124.1 | NP_956418.1 | 5 - 6 | 1961 | 97 | |
GenBank mRNA | BC053313.1 | - | 5 - 6 | 1961 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
