Assay Details
Gene Symbol: ncbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200143.1 | NP_956437.1 | 8 - 9 | 1053 | 61 | |
GenBank mRNA | BC044520.1 | - | 8 - 9 | 1053 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
