Assay Details
Gene Symbol: tubg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001320515.1 | NP_001307444.1 | 3 - 4 | 444 | 70 | |
NM_200908.2 | NP_957202.1 | 3 - 4 | 444 | 70 | ||
XM_017353659.2 | XP_017209148.1 | 3 - 4 | 451 | 70 | ||
GenBank mRNA | BC045486.1 | - | 413 | 70 | ||
BC159223.1 | - | 3 - 4 | 438 | 70 | ||
EH487910.1 | - | 3 - 4 | 452 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
