Assay Details
Gene Symbol: e2f5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197300.1 | NP_001184229.1 | 1 - 2 | 307 | 121 | |
NM_205615.2 | NP_991178.1 | 1 - 2 | 307 | 121 | ||
GenBank mRNA | AY398338.1 | - | 1 - 2 | 121 | ||
BC098598.1 | - | 1 - 2 | 121 | |||
EB911484.1 | - | 1 - 2 | 183 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
