Assay Details
Gene Symbol: cbr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205656.2 | NP_991219.2 | 5 - 6 | 630 | 111 | |
GenBank mRNA | BC065615.1 | - | 5 - 6 | 611 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
