Assay Details
Gene Symbol: nedd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213506.1 | NP_998671.1 | 12 - 13 | 1584 | 74 | |
XM_005163004.4 | XP_005163061.1 | 12 - 13 | 1617 | 74 | ||
GenBank mRNA | BC058053.1 | - | 12 - 13 | 1584 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
