Assay Details
Gene Symbol: nedd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213506.1 | NP_998671.1 | 15 - 16 | 2050 | 120 | |
XM_005163004.4 | XP_005163061.1 | 15 - 16 | 2086 | 120 | ||
GenBank mRNA | BC058053.1 | - | 15 - 16 | 2050 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
