Assay Details
Gene Symbol: nedd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213506.1 | NP_998671.1 | 8 - 9 | 1271 | 61 | |
XM_005163004.4 | XP_005163061.1 | 8 - 9 | 1304 | 61 | ||
GenBank mRNA | BC058053.1 | - | 8 - 9 | 1271 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
