Assay Details
Gene Symbol: scnm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001813.1 | NP_001001813.1 | 2 - 3 | 222 | 100 | |
GenBank mRNA | AY398412.1 | - | 2 - 3 | 222 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
