Assay Details
Gene Symbol: th2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001829.1 | NP_001001829.1 | 8 - 9 | 1030 | 133 | |
GenBank mRNA | AY620823.1 | - | 8 - 9 | 1030 | 133 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
