Assay Details
Gene Symbol: adipor1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC076349.1 | - | 5 - 6 | 673 | 81 | |
BC165131.1 | - | 3 - 4 | 341 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
