Assay Details
Gene Symbol: adipor1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001327754.1 | NP_001314683.1 | 8 - 9 | 1340 | 63 | |
GenBank mRNA | BC076349.1 | - | 7 - 8 | 802 | 63 | |
EH578188.1 | - | 1 - 2 | 664 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
