Assay Details
Gene Symbol: crebl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002702.2 | NP_001002702.2 | 3 - 4 | 438 | 102 | |
GenBank mRNA | BC081675.1 | - | 3 - 4 | 438 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
