Assay Details
Gene Symbol: ndufa7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003436.2 | NP_001003436.1 | 1 - 2 | 214 | 103 | |
GenBank mRNA | BC076312.1 | - | 1 - 2 | 63 | 103 | |
BC164444.1 | - | 1 - 2 | 86 | 103 | ||
EH500639.1 | - | 1 - 2 | 105 | 103 | ||
EH548721.1 | - | 1 - 2 | 254 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
