Assay Details
Gene Symbol: syf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003437.2 | NP_001003437.1 | 2 - 3 | 133 | 95 | |
GenBank mRNA | BC076298.1 | - | 2 - 3 | 134 | 95 | |
BC091556.1 | - | 2 - 3 | 161 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
