Assay Details
Gene Symbol: coq5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004541.1 | NP_001004541.1 | 3 - 4 | 588 | 120 | |
GenBank mRNA | BC078433.1 | - | 3 - 4 | 588 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
