Assay Details
Gene Symbol: lypd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004670.1 | NP_001004670.1 | 2 - 3 | 318 | 94 | |
XM_021478643.1 | XP_021334318.1 | 2 - 3 | 184 | 94 | ||
GenBank mRNA | BC081426.1 | - | 2 - 3 | 318 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
