Assay Details
Gene Symbol: nipa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006062.1 | NP_001006062.1 | 3 - 4 | 410 | 103 | |
GenBank mRNA | BC083262.1 | - | 3 - 4 | 410 | 103 | |
BC164079.1 | - | 3 - 4 | 289 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
