Assay Details
Gene Symbol: nap1l4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007453.1 | NP_001007454.1 | 4 - 5 | 326 | 79 | |
XM_009303210.3 | XP_009301485.1 | 4 - 5 | 403 | 79 | ||
XM_021477517.1 | XP_021333192.1 | 3 - 4 | 340 | 79 | ||
GenBank mRNA | BC085386.1 | - | 4 - 5 | 326 | 79 | |
BC164768.1 | - | 3 - 4 | 291 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
