Assay Details
Gene Symbol: cnih2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013524.1 | NP_001013542.1 | 3 - 4 | 493 | 92 | |
XM_021479171.1 | XP_021334846.1 | 5 - 6 | 92 | |||
XM_021479172.1 | XP_021334847.1 | 5 - 6 | 92 | |||
GenBank mRNA | BC090830.1 | - | 3 - 4 | 493 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
